missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Scott™ Essential™ Coreless SRB Tissue Dispenser

Catalog No. 19813851
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19813851 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19813851 Supplier Kimberly-Clark Professional™ Supplier No. 09605

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sleek and durable. Holds 2 full standard rolls of coreless tissue. When installed properly this dispenser meets the ADA Standards for Accessible Design; 28 CFR Part 36; local rules may vary.

  • Compatible with two standard coreless toilet paper rolls
  • Dispenser measures 11.0 in. x 7.63 in. x 6.0 in.
  • Manual dispenser offers easy refills and maintenance with no lock
  • Hygienic design covers both rolls
  • When properly installed, these dispensers meet the ADA Standards for Accessible Design.

Specifications

Product Type Coreless Double Roll Bathroom Tissue Dispenser
Color White
Holds Bathroom Tissue
Material Plastic
Mounting Wall Mount
No. of Compartments 2
Product Style Front-loading with Opening
Certifications/Compliance ADA Standard
For Use With (Application) Provides built-in anti-microbial protection against most common stain and odor-causing bacteria
Sterility Non-sterile