Learn More
Ansell Kimtech™ A8 Certified Lab Coats with Knit Cuffs and Collar
Breathable SMS lab coats protect from light fluid contact and feature soft knit cuffs and 3 pockets. Choose from white or blue colors.
Supplier:  Ansell ASL10030
 View more versions of this product
Catalog No. 29-635-9856
Only null left
Explore available promotions
Description
Description
  • Lab coats with knit cuffs and collar
  • Comfort and protection from light fluid contact
  • 3-layer SMS nonwoven fabric
  • Soft knit cuffs and collar
  • Mid-calf length
  • Three reinforced pockets and five snaps
  • Neck loop for convenient hanging
Specifications
Specifications
General Purpose Lab Coat
Unisex
Blue
Knit
Non-sterile
3
10030
3-ply
3-ply SMS
Small
SMS
Snaps
Limited-use
Mid-calf Length
Knit
30 to 34 in.
Hanging Neck Loop
Product Suggestions
Product Suggestions