missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Kimtech™ A8 Lab Coats, Traditional Collar

Description
- Protective three-layer white SMS fabric resistant to blood strike-through for various levels of exposure
- Feature traditional collar, knit cuffs, four snaps, two reinforced pockets, and size/name label
OSHA requires the employer to evaluate the task and type of exposure anticipated and select the appropriate clothing that prevents contamination of an employee's skin or clothing by blood or other potentially infectious materials
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | Large |
| Gender | Unisex |
| Material | SMS |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Sterility | Non-sterile |
| No. of Pockets | 2 |
| Collar Type | Traditional |
| Show More |