Learn More
Ansell A8 Ankle-High Shoe Covers with Traction Strips
Catalog No. 296359901
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296359901 X-Large
19188584 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296359901 Supplier Ansell Supplier No. 69353
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Shoe covers fit over the ankle with elastic and extend to mid-calf for extra protection from moderate fluid contact. Adhesive traction strips on the bottom help prevent slips.
  • Three layers of SMS fabric
  • Adhesive traction strips
  • For moderate fluid contamination
  • Latex free
Specifications
Size X-Large
Material SMS
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Sole Type Traction Strips
Model 69353
Thickness 3-ply
Includes Adhesive traction stripes
Resistance Fluid
Product Type Shoe Covers
Show More Show Less