Learn More
DWK Life Sciences Kimble™ KIMAX™ U-shaped drying tube, Atomizer connector tube
Catalog No. 12141414
Change view
Click to view available options
For Use With (Equipment):
17124F Burette
Catalog No. For Use With (Equipment)
12141414 17124F Burette
1 options
Catalog No. 12141414 Supplier DWK Life Sciences Supplier No. 17324F0001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement Parts - Drying tube, connecting tube, and rubber stopper for 17124F Buret
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
For Use With (Equipment) 17124F Burette
Type Drying Tube