Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim and Marking Spot
Marking spot is durable fused-on white enamel
Supplier:  DWK Life Sciences 4504216150
 View more versions of this product
Catalog No. 14956E
Only null left
Description
Description
Marking spot is durable fused-on white enamel
  • Kimax KG-33 glass
  • With sturdy lip
  • Uniform wall thickness for maximum heat transfer, chemical resistance
Specifications
Chemical Identifiers
ASTM E982, Type I
Plain Beaded Rim
Borosilicate Glass
Marking Spot
Specifications
ASTM E982, Type I
24 mL
Borosilicate Glass
16 mm
Plain Beaded Rim
150 mm
Marking Spot
Product Suggestions
Product Suggestions