Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim and Marking Spot
Marking spot is durable fused-on white enamel
Supplier: DWK Life Sciences 4504215125
Description
Marking spot is durable fused-on white enamel
- Kimax KG-33 glass
- With sturdy lip
- Uniform wall thickness for maximum heat transfer, chemical resistance
Chemical Identifiers
| ASTM E982, Type I | |
| Plain Beaded Rim | |
| Borosilicate Glass | |
| Marking Spot |
Specifications
| ASTM E982, Type I | |
| 16 mL | |
| Borosilicate Glass | |
| 15 mm |
| Plain Beaded Rim | |
| 125 mm | |
| Marking Spot |