Learn More
Kimble™ KIMAX™ Narrow Mouth Erlenmeyer Flasks
Made of quality borosilicate glass.
$269.77 - $269.77
Specifications
| Closure Material | Rubber |
|---|---|
| Description | White Ceramic Enamel capacity scale; Top style: Reinforced and tooled with rounded finish beaded |
| Material | Borosilicate Glass |
| Mouth Style | Narrow |
Description
- Perfectly suited for general laboratory use
- White ceramic enamel marking spot and a capacity scale for ease of reading
- Sizes 25, 50, and 125mL are ideal for microscale measurements
Specifications
| Rubber | |
| White Ceramic Enamel capacity scale; Top style: Reinforced and tooled with rounded finish beaded | |
| Borosilicate Glass | |
| Narrow |