Learn More
DWK Life Sciences Kimble™ KIMAX™ GL 45 Media Bottle Starter Pack
An assortment of popularly sized media bottles from our 14395 series, which is ideal for general laboratory use including mixing, storing, or transporting culture media, chemicals, or solvents.
Supplier:  DWK Life Sciences 1439501DUP
Catalog No. S15001
Only null left
Explore available promotions
Description
Description
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied with linerless GL 45 screw thread caps
  • Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
  • The pack consists of two 100 mL, three 250 mL, three 500 mL, and two 1000 mL bottles
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Media Bottle Starter Pack
Product Suggestions
Product Suggestions