Learn More
DWK Life Sciences Kimble™ KIMAX™ Desiccator Glass Stopcock Valve and Plug with Neoprene Bushing
Catalog No. 12141422
Change view
Click to view available options
For Use With (Equipment):
Borosilicate Desiccators 21200-200 and 21200-250
Catalog No. For Use With (Equipment)
12141422 Borosilicate Desiccators 21200-200 and 21200-250
1 options
Catalog No. 12141422 Supplier DWK Life Sciences Supplier No. 211752
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement Parts - Glass Stopcock for Borosilicate Desiccators 21200-200 and 21200-250
  • Simplified stopcock design minimizes chipping or marring of critically ground finishes during cleaning and assembly
  • Valve is secured to the desiccator lid by a neoprene grommet located in the knob of the cover
  • Stem is tooled with a hose connection to accept 5/16 in. ID flexible tubing
  • All glass apparatus manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Material Glass
For Use With (Application) Vacuum desiccation
For Use With (Equipment) Borosilicate Desiccators 21200-200 and 21200-250
Includes Neoprene bushing
Product Type Valve Stopcock