missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Class B Cylinders, Starter Pack

Catalog No. 03340197
Change view
Click to view available options
Tolerance:
±0.1 mL (at 10 mL), ±0.3 mL (at 25 mL), ±0.4 mL (at 50 mL), ±0.6 mL (at 100 mL), ±1.4 mL (at 250 mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tolerance
03340197 ±0.1 mL (at 10 mL), ±0.3 mL (at 25 mL), ±0.4 mL (at 50 mL), ±0.6 mL (at 100 mL), ±1.4 mL (at 250 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03340197 Supplier DWK Life Sciences Supplier No. 2002401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

To deliver; Class B

  • One each 10, 25, 50, 100, 250mL
  • Single metric scale with bumper

Specifications

Material Borosilicate Glass
Capacity (Metric) 10 mL, 100 mL, 25 mL, 250 mL, 50 mL
For Use With (Application) Start Up Labs and Customers Who Need a Variety of Cylinders But Have Limited Lab Space or Glassware Needs
Scale Single
Graduations 0.2 mL (10 mL), 0.5 mL (25 mL), 1 mL (100 mL), 1 mL (50 mL), 2 mL (250 mL)
Subdivision 0.2 to 2mL
Calibration To Deliver
Base Type Round
For Use With (Equipment) Bumper
Shape Round
Class Class B
Height (Metric) 190 mm (50 mL), 220 mm (250 mL), 255 mm (100 mL), 135 mm (10 mL), 140 mm (25 mL)
Certifications/Compliance ASTM E1272, ASTM E438, USP Type I
Tolerance ±0.1 mL (at 10 mL), ±0.3 mL (at 25 mL), ±0.4 mL (at 50 mL), ±0.6 mL (at 100 mL), ±1.4 mL (at 250 mL)
Product Type Graduated Cylinder Set
Show More Show Less