missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences KIMBLE™ KIMAX™ Brand Replacement Base for 25mL Educational-Grade Graduated Cylinders
Description
- Genuine replacement base for KIMAX 25mL educational-grade graduated cylinders
- Compatible with KIMAX cylinders featuring a single blue metric scale
- Sold as replacement part to extend the service life of existing cylinders
- Case quantity: 6
Specifications
Specifications
| Material | Borosilicate Glass |
| Capacity (Metric) | 25 mL |
| For Use With (Application) | 25 mL Cylinder |
| Scale | Single metric scale |
| Product Type | Cylinder Base |