missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Brand Pipet Dropping Bottle

Medium length pipet dropper

Supplier:  DWK Life Sciences 15035125

 View more versions of this product

Catalog No. 02-990-2C


Only null left

Description

Description

Medium length pipet dropper

  • Rubber nipple
  • KG-33 ASTM 438 Type 1 Class A Glass
  • Dropper Capacity: 1.0mL

Specifications

Specifications

4.22 oz., 125 mL
4.22 oz.
Borosilicate Glass
1 mL
Pipet Dropping Bottle
55 mm
125 mL
2.16 in.
19/22
150 mm
5.9 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions