missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Automatic Burets with Bottles

Catalog No. p-47145
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
038482A 10 mL
038482B 25 mL
038482C 50 mL
038482D 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 038482A Supplier DWK Life Sciences Supplier No. 17124F10
Only null left

Class A

standard taper joints fit into reagent bottles with standard taper necks.

  • Constructed with overflow tubes that return directly to reservoir
  • For air intake, overflow cups have hose connection for attaching drying tube; second drying tube provided for use between rubber bulb and reservoir
  • Precision bore tubing permanently marked with individual serial number
  • Durable black ceramic enamel scale
  • Inset shows top configuration

Certifications

A certificate is provided to indicate instrument traceability to standards provided by NIST™.

Specifications

Capacity (Metric) 10 mL
Length (Metric) 765 mm
Material Borosilicate Glass
Graduations 0.05 mL
For Use With (Application) Volumetric Analysis
Length (English) 30.1 in.
Includes Reservoir Bottle, U-Shaped Drying Tube, Vented Connecting Tube, Rubber Squeeze Bulb, No. 1 Single-Holed Rubber Stopper, PTFE Stopcock Plug and 1/4 In. I.D. Rubber Tubing, Durable Black Ceramic Enamel Scale
Scale Black
Standard Taper Joint 29
Stopcock Material PTFE
Tolerance ±0.02 mL
Volume (Metric) Reservoir 1000 mL
Type Automatic Buret
Class Class A
Certifications/Compliance ASTM E438
Show More Show Less