missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Kick-Net

Catalog No. S85005P
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S85005P 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S85005P Supplier LaMotte™ Supplier No. 0021P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Strong net designed for rapid bioassessments.

  • Use this net for performing USEPA Rapid Bioassessment Protocols for benthic macroinvertebrates
  • 1 x 1m2, 500μm, tan net
  • Hemmed sides hold poles
  • Reinforced bottom seam helps anchoring

Specifications

Grade 3+
Product Type Professional Kick Net with Poles
Includes Poles
For Use With (Application) Performing USEPA rapid bioassessment protocols for benthic macroinvertebrates