missing translation for 'onlineSavingsMsg'
Learn More

Ansell KGA20 Hooded Coveralls

Catalog No. 17101167
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17101167 Medium
17101168 Large
17101169 X-Large
17101170 2X-Large
17101171 3X-Large
17101172 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17101167 Supplier Ansell Supplier No. 68971

Coveralls with attached hood provide lightweight protection against heavy dust and limited splashes.

  • Comfortable, lightweight, SMS fabric is breathable and allows moisture to escape
  • Hooded, Elastic Wrist and Ankle Design
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Dust, Liquid
Certifications/Compliance ANSI/ISEA 101-1996
Certifications/Compliance (Europe) CE Category I PPE
Latex Free Yes
Model KleenGuard™ KGA20
For Use With (Application) Agriculture, Food Processing, General Maintenances, Laboratories
Show More Show Less