Learn More
Ansell KleenGuard™ KGA10 Lab Coats
Highly breathable spunbond fabric lab coats are CE Category I PPE and are free from natural rubber latex and silicone materials.
Supplier:  Ansell 67317
 View more versions of this product
Catalog No. 17-101-160
Only null left
Explore available promotions
Description
Description
  • For non-hazardous particulate protection
  • Lightweight and highly breathable spunbond fabric allows moisture to escape for maximum comfort
  • 4-Snap Closure, Elastic Wrists, No Pockets
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program
Specifications
Specifications
General Purpose Lab Coat
Unisex
Snaps
Single-use
None
KGA10
2X-Large
White
Elastic
Non-sterile
Yes
Stitched
Product Suggestions
Product Suggestions