missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KleenGuard™ KGA10 Lab Coats
Highly breathable spunbond fabric lab coats are CE Category I PPE and are free from natural rubber latex and silicone materials.
Supplier: Ansell KC67316
Description
- For non-hazardous particulate protection
- Lightweight and highly breathable spunbond fabric allows moisture to escape for maximum comfort
- 4-Snap Closure, Elastic Wrists, No Pockets
- Not made with natural rubber latex or silicone
- Recyclable with the RightCycle™ Program
Specifications
| General Purpose Lab Coat | |
| Unisex | |
| Snaps | |
| Single-use | |
| None | |
| KGA10 |
| X-Large | |
| White | |
| Elastic | |
| Non-sterile | |
| Yes | |
| Stitched |