Learn More
Ansell KleenGuard™ KGA10 Lab Coats
GREENER_CHOICE
Catalog No. 17101158
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17101158 Large
17101159 X-Large
17101160 2X-Large
17101161 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17101158 Supplier Ansell Supplier No. 67315
Highly breathable spunbond fabric lab coats are CE Category I PPE and are free from natural rubber latex and silicone materials.
  • For non-hazardous particulate protection
  • Lightweight and highly breathable spunbond fabric allows moisture to escape for maximum comfort
  • 4-Snap Closure, Elastic Wrists, No Pockets
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Latex Free Yes
Model KGA10
Seam Type Stitched
Show More Show Less