missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety White SMS Lab Coats, Snap Front, Elastic Wrists

Catalog No. 19168639
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19168639 X-Large
19168636 Small
19168637 Medium
19168638 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19168639 Supplier Keystone Safety Supplier No. LC0WESMSXLWHITE

Latex free SMS lab coats provide barrier protection and are comfortable to wear.

  • Latex free, 3-layer SMS fabric
  • Barrier against water based liquids and light chemical splash
  • Single collar
  • Strong, high tensile strength
  • Comfortable, lightweight, breathable
  • Serged seams
  • Meets ANSI 101-1996 and Federal Flammability ACT 16 CFR 1610.
  • Multi use for cleanroom, pharmaceutical, food, janitorial, cleaning, maintenance
  • Keystone LC0-WE-SMS-WHITE SMS Lab Coat, No Pockets, Elastic Wrists, Snap Front
  • White, Pack of 30

Specifications

Product Type Controlled Environments Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Collar Type Single
Latex Free Yes
Model LC0-WE-SMS
Seam Type Serged
Certifications/Compliance ANSI 101-1996
For Use With (Application) Janitorial, Manufacturing, Food Service, Industrial, Medical, Cleanroom, Pharmaceutical
Length (English) 40 in.
Length (Metric) 101.6 cm
Weight (English) 1.4 oz.
Autoclavable Not Autoclavable
No. per Pack 30/Pk.
Show More Show Less