missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keystone Safety Nylon Beard Nets, 1/16 in. Holes
Description
- Beard net
- Soft heavyweight nylon, 1/16 in. hole
- Honeycomb style
- Flame-resistant
- Available in large, x-large sizes
- Available in white or blue color
Specifications
Specifications
| Size | Large |
| Material | Nylon |
| Color | White |
| Disposable | Single-use |
| For Use With (Application) | Arc Flash Industry, Electrical |
| Product Type | General Purpose Beard Cover |