missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety Heavy Duty Polypropylene Cleanroom Lab Coats

Catalog No. 19068535
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19068535 4X-Large
19068431 Medium
19068432 Large
19068434 2X-Large
19068433 X-Large
19068530 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19068535 Supplier Keystone Safety Supplier No. LC0WENWHD4XLWHITE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lightweight, latex-free lab coats are made from high quality materials and compatible in Class ISO7 cleanroom environments.

Protect workers against dry particulates, dirt and grime

  • Non-woven, nonlinting, spunbond heavyweight polypropylene construction
  • Serged seams offer high tensile strength
  • Latex-free
  • Disposable
  • Elastic wrists, snap front, single collar, no pockets
  • Available in six sizes
  • Flame resistant

Ideal for Class 10,000 environments and above. Commonly used in electronic, medical device and pharmaceutical industries where operator garment control is a concern.

Specifications

Product Type Controlled Environments Lab Coat
Size 4X-Large
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Collar Type Single Ply
Seam Type Serged
For Use With (Application) Electronics, Medical Device, Pharmaceutical
Cleanroom Class Class 10,000 (ISO 7)
Show More Show Less