missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety KeyGuard Microporous Lab Coats

Catalog No. 19070104
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19070104 2X-Large
19070100 Small
19070101 Medium
19070102 Large
19070103 X-Large
19070105 3X-Large
19070106 4X-Large
19070107 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19070104 Supplier Keystone Safety Supplier No. LCO WE KG 2XL
Only null left

Latex free microporous lab coats are a cost savings alternative to Tyvek and provide contamination control in a variety of environments including cleanrooms.

  • Non-linting
  • Microporous
  • Latex free
  • Cost savings alternative to Tyvek brand
  • Individually inspected for stitching, sizing, material and workmanship
  • Ideal for cleanroom environments including electronic, medical device, and pharmaceutical industries where garment control is a concern

Specifications

Product Type Controlled Environments Lab Coat
Size 2X-Large
Gender Unisex
Material Microporous Film
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Latex Free Yes
Model LCO-WE-KG
Seam Type Serged
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
No. per Pack 30/Pk.
Construction Keyguard
Show More Show Less