missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Keyed Alike Safety Lock with Steel Shackle
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient and cost-effective Safety Lockout Padlocks from Brady.
Supplier: Brady™ SDPLRED38STKA12
Description
- Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program.
- Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
- Nylon padlocks with steel shackles are lightweight, durable and ideal for general lockout applications.
- Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
- Record and retain engraved key codes to streamline replacement of lost keys.
Chemical Identifiers
| Padlock | |
| Nylon | |
| 6.35 mm | |
| Ideal for electrical contractors, maintenance, electricians | |
| Steel |
| Keyed Alike | |
| 0.25 in. | |
| 0.82 x 1.5 x 1.77 in. | |
| 38.10 mm | |
| 6 |
Specifications
| Padlock | |
| Red | |
| 0.25 in. | |
| 0.82 x 1.5 x 1.77 in. | |
| 38.10 mm | |
| Steel | |
| 12 Pk. |
| Keyed Alike | |
| Nylon | |
| 6.35 mm | |
| Ideal for electrical contractors, maintenance, electricians | |
| (12) Padlocks, (12) Keys, (12) English Labels, (12) French Labels, (12) Spanish Labels | |
| 6 |