missing translation for 'onlineSavingsMsg'
Learn More

Ketotifen Fumarate 98.0+%, TCI America™

Catalog No. p-7131758
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00481G 1 g
K00485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00481G Supplier TCI America Supplier No. K00481G

Chemical Identifiers

CAS 34580-14-8
Molecular Formula C23H23NO5S
Molecular Weight (g/mol) 425.499
MDL Number MFCD00079394
InChI Key YNQQEYBLVYAWNX-WLHGVMLRSA-N
Synonym ketotifen fumarate, zaditen, ketotifen hydrogen fumarate, alaway, unii-hbd503woro, hc 20,511 fumarate, ketotifen fumarate usan:jan, ketotifen fumarate salt, hbd503woro
PubChem CID 5282408
IUPAC Name (E)-but-2-enedioic acid;10-(1-methylpiperidin-4-ylidene)-5H-benzo[1,2]cyclohepta[3,4-b]thiophen-4-one
SMILES CN1CCC(=C2C3=C(C(=O)CC4=CC=CC=C42)SC=C3)CC1.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 1 g
Formula Weight 425.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ketotifen Fumarate
RTECSNumber DE8260000
TSCA No