missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketorolac Tromethamine 98.0+%, TCI America™
$100.11 - $363.85
Chemical Identifiers
| CAS | 74103-07-4 |
|---|---|
| Molecular Formula | C19H24N2O6 |
| Molecular Weight (g/mol) | 376.41 |
| MDL Number | MFCD00887595 |
| InChI Key | BWHLPLXXIDYSNW-UHFFFAOYNA-N |
| Synonym | Ketorolac Tris Salt, (+/-)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid Tris Salt |
| PubChem CID | 84003 |
| ChEBI | CHEBI:6130 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid |
| SMILES | NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 74103-07-4 | |
| 376.41 | |
| BWHLPLXXIDYSNW-UHFFFAOYNA-N | |
| 84003 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid |
| C19H24N2O6 | |
| MFCD00887595 | |
| Ketorolac Tris Salt, (+/-)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid Tris Salt | |
| CHEBI:6130 | |
| NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1 |
Specifications
| 74103-07-4 | |
| White | |
| MFCD00887595 | |
| 2811 | |
| BWHLPLXXIDYSNW-UHFFFAOYNA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid | |
| 84003 | |
| 376.41 | |
| Crystalline Powder |
| 167°C | |
| C19H24N2O6 | |
| 1 g | |
| Ketorolac Tris Salt, (+/-)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid Tris Salt | |
| NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1 | |
| 376.41 | |
| CHEBI:6130 | |
| ≥98.0% (HPLC,T) | |
| Ketorolac Tromethamine |
Safety and Handling
RTECSNumber : UY7759900
TSCA : No