missing translation for 'onlineSavingsMsg'
Learn More

Ketorolac Tromethamine 98.0+%, TCI America™

Catalog No. K00535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00535G 5 g
K00531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00535G Supplier TCI America Supplier No. K00535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74103-07-4
Molecular Formula C19H24N2O6
Molecular Weight (g/mol) 376.41
MDL Number MFCD00887595
InChI Key BWHLPLXXIDYSNW-UHFFFAOYNA-N
Synonym Ketorolac Tris Salt, (+/-)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid Tris Salt
PubChem CID 84003
ChEBI CHEBI:6130
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
SMILES NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1

Specifications

Melting Point 167°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 376.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ketorolac Tromethamine
RTECSNumber UY7759900
TSCA No