missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketoketal, 97%, Thermo Scientific™
$77.84 - $268.33
Chemical Identifiers
| CAS | 56309-94-5 |
|---|---|
| Molecular Formula | C14H22O3 |
| Molecular Weight (g/mol) | 238.327 |
| MDL Number | MFCD00809704 |
| InChI Key | ZNWLFTSPNBLXGL-UHFFFAOYSA-N |
| Synonym | ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl |
| PubChem CID | 810913 |
| IUPAC Name | 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one |
| SMILES | C1CC(=O)CCC1C2CCC3(CC2)OCCO3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5506906
|
Thermo Scientific Chemicals
H5506906 |
5 g |
N/A
|
|
|||||
|
AAH5506914
|
Thermo Scientific Chemicals
H5506914 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsKetoketal is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 56309-94-5 | |
| 238.327 | |
| ZNWLFTSPNBLXGL-UHFFFAOYSA-N | |
| 810913 | |
| C1CC(=O)CCC1C2CCC3(CC2)OCCO3 |
| C14H22O3 | |
| MFCD00809704 | |
| ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl | |
| 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one |
Specifications
| 56309-94-5 | |
| MFCD00809704 | |
| ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl | |
| ZNWLFTSPNBLXGL-UHFFFAOYSA-N | |
| 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one | |
| 810913 | |
| 97% |
| C14H22O3 | |
| 5 g | |
| Insoluble in water. | |
| C1CC(=O)CCC1C2CCC3(CC2)OCCO3 | |
| 238.327 | |
| 238.33 | |
| Ketoketal |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
TSCA : No
Recommended Storage : Ambient temperatures