missing translation for 'onlineSavingsMsg'
Learn More

alpha-Ketoglutaric acid sodium salt, 98%

Catalog No. AC439351000
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439351000 100 g
AC439350050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439351000 Supplier Thermo Scientific Chemicals Supplier No. 439351000
Only null left

CAS: 22202-68-2 | C5H5NaO5 | 168.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22202-68-2
Molecular Formula C5H5NaO5
Molecular Weight (g/mol) 168.08
InChI Key MOTOGHHLNTXPTI-UHFFFAOYSA-M
Synonym sodium hydrogen 2-oxoglutarate, pentanedioic acid, 2-oxo-, sodium salt, pentanedioic acid, 2-oxo-, sodium salt 1:?, 2-ketoglutaric acid monosodium salt, 2-oxoglutaric acid, sodium salt, glutaric acid, 2-oxo-, sodium salt, alpha-ketoglutaric acid, sodium salt, 2-oxoglutaric acid 1-sodium salt, sodium 4-carboxy-4-oxobutanoate
PubChem CID 23672314
IUPAC Name sodium;5-hydroxy-4,5-dioxopentanoate
SMILES C(CC(=O)[O-])C(=O)C(=O)O.[Na+]

Specifications

Color White
CAS Min % 97.5
CAS Max % 100.0
pH 3.6 to 4.0 (0.5% aq. soln.)
Physical Form Powder
Quantity 100 g
Assay Percent Range 98%
Linear Formula HO2CCH2CH2C(=O)CO2Na
Formula Weight 168.08
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 1.0% max.
Chemical Name or Material α-Ketoglutaric acid sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 244-836-6

RUO – Research Use Only