missing translation for 'onlineSavingsMsg'
Learn More

α-Ketoglutaric acid disodium salt hydrate, ≥95%, MilliporeSigma™ Supelco™

Catalog No. 111013872
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013872 1 kg
111013871 5 g
111013870 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111013872 Supplier Merck Emd Millipore Supplier No. K37521KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Chemical Identifiers

MDL Number MFCD00150702
Synonym 2-Oxopentanedioic acid disodium salt hydrate; Sodium 2-oxoglutarate dibasic hydrate

Specifications

CAS 305-72-6
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C5H4O5 Na2· xH2 O
Quantity 1 kg
Formula Weight 190.06 (anhydrous basis)
Percent Purity ≥95%
Physical Form Neat