missing translation for 'onlineSavingsMsg'
Learn More

α-Ketoglutaric Acid Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022687
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022687 10 mg
111022688 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022687 Supplier Merck Emd Millipore Supplier No. 6123410MG

α-Ketoglutaric acid, an α-ketoacid, is identified as a potential, renal cell carcinoma (RCC) biomarker via urinary metabolomic profiling. It plays an important role in living beings as an angiogenesis suppressor.

Specifications

CAS 328-50-7
Flash Point Not applicable
Molecular Formula C5H6O5
Linear Formula HOOCCH2CH2COCOOH
Quantity 10 mg
MDL Number MFCD00004165
UN Number NONH for all modes of transport
Synonym 2-Oxoglutaric Acid; 2-Oxopentanedioic Acid
Formula Weight 146.10
Percent Purity 99% to 101% (T)
Grade Analytical Standard
Show More Show Less