missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole 98.0+%, TCI America™

Catalog No. p-7131756
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
K00451G 1 g
K00455G 5 g
2 options

Catalog No. K00451G

Supplier: TCI America K00451G

Only null left

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
MDL Number MFCD00058579
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym ketoconazole
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 531.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ketoconazole
RTECSNumber TK7912300
TSCA No
Recommended Storage Refrigerator