missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019579
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019579 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019579 Supplier Merck Emd Millipore Supplier No. PHR13851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Ketoconazole is an imidazole antifungal agent administered through topical or oral means. It is used for the treatment of chronic mucocutaneous candidiasis, fungal infections of the gastro-intestinal tract, dermatophyte infections, systemic infections, and fungal infections in immuno-compromised patients. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 65277-42-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C26H28 Cl2N4O4
MDL Number MFCD00058579
Synonym Ketoconazole; (+/-)-cis-1-Acetyl-4-(4-[(2-[2,4-dichlorophenyl]-2-[1 H-imidazol-1-ylmethyl]-1,3-dioxolan-4-yl)-methoxy]phenyl)piperazine
Type Certified Reference Material
Quantity 1 g
Formula Weight 531.43
Grade Certified Reference Material