missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, 98%

Potent inhibitor of cytochrome P450c17 | CAS: 65277-42-1 | C26H28Cl2N4O4 | 534.452 g/mol

Supplier:  Thermo Scientific Chemicals J6336714

 View more versions of this product

Catalog No. AAJ6336714


Only null left
Explore available promotions

Description

Description

Ketoconazole is used to treat candidiasis, chronic mucocutaneous candidiasis, oral thrush, candiduria, blastomycosis, coccidioidomycosis, histoplasmosis, chromomycosis, and paracoccidioidomycosis. Ketoconazole is an antifungal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ketoconazole is used to treat candidiasis, chronic mucocutaneous candidiasis, oral thrush, candiduria, blastomycosis, coccidioidomycosis, histoplasmosis, chromomycosis, and paracoccidioidomycosis. Ketoconazole is an antifungal agent.

Solubility
Soluble in DMSO, ethanol, chloroform, water, and methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Ketoconazole
White
MFCD00058579
UN2811
ketoconazole
XMAYWYJOQHXEEK-SIULDFEJSA-N
2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
76973198
98%
65277-42-1
C26H28Cl2N4O4
25 g
14,5302
Soluble in DMSO,ethanol,chloroform,water,and methanol.
CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl
534.452
531.43
Powder

Chemical Identifiers

65277-42-1
534.452
XMAYWYJOQHXEEK-SIULDFEJSA-N
76973198
CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl
C26H28Cl2N4O4
MFCD00058579
ketoconazole
2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H360-H373
Toxic if swallowed.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c

H301-H360F-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 265-667-4

RTECSNumber : TK7912300

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only