missing translation for 'onlineSavingsMsg'
Learn More

Ketanserin 98.0+%, TCI America™

Catalog No. k0051200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K0051200MG 200 mg
K00511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K0051200MG Supplier TCI America Supplier No. K0051200MG

Chemical Identifiers

CAS 74050-98-9
Molecular Formula C22H22FN3O3
Molecular Weight (g/mol) 395.434
MDL Number MFCD00083392
InChI Key FPCCSQOGAWCVBH-UHFFFAOYSA-N
Synonym 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2,4(1H,3H)-quinazolinedione
PubChem CID 3822
ChEBI CHEBI:6123
IUPAC Name 3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
SMILES C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 200 mg
Formula Weight 395.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ketanserin
RTECSNumber VA1411190
TSCA No
Recommended Storage Refrigerator