missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kerosene, Reagents
Specifications
Specifications
| Boiling Range | 175°C to 325°C |
| Melting Point | -48°C to -26°C |
| Color | Red |
| Boiling Point | 175°C |
| Melting Range | -48°C to -26°C |
| Packaging | Glass Bottle |
| Quantity | 1 L |
| Specific Gravity | 0.767 |
| Grade | Technical |
| Physical Form | Liquid |
| Show More |