missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Reagent Grade, LabChem™

Catalog No. p-5379513
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC158502 1 L Amber Glass
LC158504 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC158502 Supplier LabChem Supplier No. LC158502
Only null left

Chemical Identifiers

CAS 64742-48-9

Specifications

CAS Max % 100
Melting Point -20°C
Color Colorless
Density 769kg/m3
Flash Point 61°C
Odor Odorless
Packaging Amber Glass
Quantity 1 L
UN Number UN1223
Grade Reagent
Physical Form Liquid
Chemical Name or Material Kerosene
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
If swallowed: Iimmediately call a poison center/doctor.
Do not induce vomiting.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature