missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Reagent Grade, LabChem™

Catalog No. LC158504
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC158504 4 L Amber Glass
LC158502 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC158504 Supplier LabChem Supplier No. LC158504

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64742-48-9

Specifications

CAS Max % 100
Melting Point -20°C
Color Colorless
Density 769kg/m3
Flash Point 61°C
Odor Odorless
Packaging Amber Glass
Quantity 4 L
UN Number UN1223
Grade Reagent
Physical Form Liquid
Chemical Name or Material Kerosene
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Recommended Storage Room Temperature