missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Reagent grade, MilliporeSigma™

Catalog No. MKX002020
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MKX002020 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MKX002020 Supplier Technidata Supplier No. KX002020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64742-48-9
Synonym Mixture of hydrocarbons, Hydrotreated heavy naphtha

Specifications

Quantity 200 L
Grade Reagent
Physical Form Liquid
Chemical Name or Material Kerosene, Mixture of hydrocarbons. Hydrotreated heavy naphtha.
Recommended Storage Room Temperature