missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kerosene, Petroleum Distillate, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Range | 151° to 301°C |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.