missing translation for 'onlineSavingsMsg'
Learn More

Kerosene - Odorless, Ricca Chemical

Catalog No. RSOK0100500
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOK0100500 500 mL Amber Glass Bottle
RSOK01001C 1 L Amber Glass Bottle
RSOK01004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RSOK0100500 Supplier Ricca Chemical Company Supplier No. RSOK0100500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8008-20-6

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Boiling Point 175°C to 325°C
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 100%
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Kerosene (Odorless), Ricca Chemical
Recommended Storage 15-30°C