missing translation for 'onlineSavingsMsg'
Learn More

Kerosene (Odorless), Fisher Chemical™

Catalog No. p-152392
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
K104 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K104 Supplier Thermo Fisher Scientific Supplier No. K10 4 KEROS
Only null left

Odorless

Chemical Identifiers

CAS 64742-47-8
MDL Number MFCD00135561

Specifications

Melting Point -17.8°C
Color Yellow
Density 4.5 (Air = 1.0)
Boiling Point 151.1°C
Flash Point 71°C
Odor Odorless
Assay Percent Range >99%
Packaging Glass Bottle
Quantity 4 L
UN Number 1223
Vapor Pressure 5mmHg at 38°C
Physical Form Liquid
Chemical Name or Material Petroleum distillates,hydrotreated light, Odorless
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Wear personal protective equipment. Ensure adequate ventilation. Keep away from open flames, hot surfaces and sources of ignition. Do not breathe vapors or spray mist. Do not ingest. Avoid contact with skin and eyes. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:2
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid