missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, MilliporeSigma™

$352.89 - $352.89

Chemical Identifiers

CAS 64742-48-9
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
50-947-992
View Documents
MilliporeSigma
KX00201
4 L Glass Bottle, Pre-Packed
Each for $352.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

64742-48-9

Specifications

64742-48-9
54°C
4 L
180°C to 188°C
Glass Bottle, Pre-Packed
0.75 hPa (20°C)
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature

SDS
missing translation for 'documents'

missing translation for 'documents'