missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, low odor

Catalog No. p-7046470
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14479AP 500 mL
AAL144790F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14479AP Supplier Thermo Scientific Chemicals Supplier No. L14479AP
Only null left

CAS: 64742-47-8

Kerosene is ideal for dilution of oils and other organic fluids for determination of metal traces. It is used in paints & coatings, consumer products, printing inks and agricultural chemical applications. It is also used in food contact applications such as aluminum rolling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Kerosene is ideal for dilution of oils and other organic fluids for determination of metal traces. It is used in paints & coatings, consumer products, printing inks and agricultural chemical applications. It is also used in food contact applications such as aluminum rolling.

Solubility
Soluble in water (0.02 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Chemical Identifiers

CAS 64742-47-8
MDL Number MFCD00135561
Synonym Kerosine

Specifications

Density 0.8
Boiling Point 200°C to 250°C
Flash Point 76°C (169°F)
Refractive Index 1.444
Quantity 500 mL
Merck Index 14,5294
Solubility Information Soluble in water (0.02g/L at 20°C).
Chemical Name or Material Kerosene, low odor
Hazard Category H227-H304
Hazard Statement GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
Precautionary Statement P210-P280-P301+P310-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KEROSENE
EINECSNumber 265-149-8
RTECSNumber OA5500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only