missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Low Odor Deodorized, Reagents

Catalog No. 01334875
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334875 10 L
01334874 1 L
01334873 200 L
01334872 20 L
01334871 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01334875 Supplier Labchem Inc Supplier No. CK19000010E6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kerosene, Low Odor Deodorized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64742-48-9
Molecular Formula C12H26-C15H32
Synonym Paraffin oil

Specifications

Color Colorless to Slight Yellow
Boiling Point 175°C
Packaging Steel Pail
Quantity 10 L
Specific Gravity 0.767
Grade Reagent
Physical Form Liquid
Chemical Name or Material Kerosene, Low Odor Deodorized