missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Keratinocyte Starter Kit

Catalog No. 12568010
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12568010 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12568010 Supplier Gibco™ Supplier No. 12568010

Keratinocyte Starter Kit

The Keratinocyte Starter Kit includes Defined Keratinocyte-SFM Medium, growth supplement, and Primary Human Keratinocytes.

Specifications

Cell Type Keratinocyte Cells, Epithelial Cells
Product Type Keratinocyte Starter Kit
Serum Level Serum-free
Quantity 1 Kit
Content And Storage Store in freezer (-5 to -30°C).
Culture Type Mammalian Cell Culture

For Research Use Only. Not for use in diagnostic procedures.