missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Keratinocyte-SFM (1X), with L-glutamine, without calcium chloride
GREENER_CHOICE

Catalog No. 10725018
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10725018 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10725018 Supplier Gibco™ Supplier No. 10725018

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Keratinocyte-SFM (1X), with L-glutamine, without calcium chloride

With: L-Glutamine

Without: Calcium Chloride.

Specifications

Cell Type Keratinocyte Cells, Epithelial Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Without Additives No Calcium Chloride
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C).
Culture Type Mammalian Cell Culture
Show More Show Less
Which serum-free media can be used to culture mouse keratinocytes?

The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.

For how long can I store Keratinocyte-SFM at 4 degrees C?

The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C

For Research Use Only. Not for use in diagnostic procedures.