missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keratin, Powder, Spectrum™ Chemical
K3030
$188.88 - $188.88
Chemical Identifiers
| CAS | 69430-36-0 |
|---|---|
| Molecular Formula | C2H2BrClO2 |
| Molecular Weight (g/mol) | 173.39 |
| InChI Key | GEHJBWKLJVFKPS-UHFFFAOYNA-N |
| IUPAC Name | 2-bromo-2-chloroacetic acid |
| SMILES | OC(=O)C(Cl)Br |
Description
Spectrum™ Chemical Keratin, Powder is the key material that makes up the human bodyâs out layer of skin. It is also the key element of hair and nails. It is considered to be similar to chitin. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 69430-36-0 | |
| 173.39 | |
| 2-bromo-2-chloroacetic acid |
| C2H2BrClO2 | |
| GEHJBWKLJVFKPS-UHFFFAOYNA-N | |
| OC(=O)C(Cl)Br |
Specifications
| 69430-36-0 | |
| Amber Glass Bottle | |
| 5 g | |
| OC(=O)C(Cl)Br | |
| 173.39 |
| 100% | |
| C2H2BrClO2 | |
| GEHJBWKLJVFKPS-UHFFFAOYNA-N | |
| 2-bromo-2-chloroacetic acid | |
| Reagent |