Learn More
StatLab KeraSoft Decalcifier
Catalog No. 21010246
Change view
Click to view available options
Form:
Solution
Catalog No. Form
21010246 Solution
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21010246 Supplier StatLab Supplier No. 1037
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Harmful if swallowed, in contact with skin, or if inhaled
KeraSoft Decalcifier increases overall efficiency in histological procedures.
  • Enhances tissue rehydration for improved cutting precision
  • Prevents tissue fracturing during sectioning
  • Optimizes staining outcomes for more precise results
Specifications
Product Type Decalcifying Solution
Quantity 500 mL
Solution Type Decalcifier
Form Solution