missing translation for 'onlineSavingsMsg'
Learn More

Kemtec™ Properties of Lipids Kit

Kit helps students explore essential nature of lipids.

Supplier:  Kemtec KMT8154

Catalog No. S25810

Only null left
Explore available promotions

Includes:
  • nstructor Manual
  • Student Data Sheets
  • MSDS
  • Petroleum Ether Solution
  • Alcoholic KOH Solution
  • HCI Solution
  • Olive Oil
  • Detergent Solution
  • Soap Solution
  • Chloroform Solution
  • Iodine Solution
  • Methanol Solution
  • Gylcerol Solution
  • Solid Fat
  • Bile Salts

Requires : Fume hood, water bath, personal protective equipment, test tubes and watch glasses

Description

Description

The tests in this kit demonstrate the properties of lipids, teach students how to identify lipids, show students how to dissolve lipids in polar substances and allow students to distinguish between saturated and unsaturated lipids.

Experiments

  • Solubility of Lipids
  • Emulsification of Lipids
  • Grease Spot Test for Fats
  • Saponification of Fats
  • Iodine Test for Absorption
  • For grades 10 to 14
  • Class size: 24 students

Order Info

Class size: 24 students working in groups of three.

Specifications

Specifications

10 to 14
Demonstrate the essential properties of lipids and enable students to detect the same
Instructor manual, MSDS, assessment manual, student data sheets, petroleum Ether solution, Alcoholic KOH solution, HCI solution, olive oil, detergent solution, soap solution, Chloroform solution, Iodine solution, Methanol solution, Gylcerol solution, solid fat and bile salts
Properties of Lipids Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions